4-(N-Isobutanoylsulphamoyl)benzeneboronic acid

APOSOR11587-250MGEA 0 EUR
APOSOR11587-250MG APOSOR11587-1G
4-(N-Isobutanoylsulphamoyl)benzeneboronic acid
4-(N-Isobutanoylsulphamoyl)benzeneboronic acid
Kaava: C₁₀H₁₄BNO₅S
MW: 271,098 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD09800863
CAS-numero: 957120-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR