2-Methyl-3-[(methylamino)methyl]furan 95%

Vaara

APOSOR12288-1GEA 0 EUR
APOSOR12288-1G APOSOR12288-250MG
2-Methyl-3-[(methylamino)methyl]furan 95%
2-Methyl-3-[(methylamino)methyl]furan
Kaava: C₇H₁₁NO
MW: 125,168 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09702377
CAS-numero: 916766-85-3
UN: 2922
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR