4-(1,3-Dithiolan-2-yl)-2-methoxy-6-nitrophenol

APOSOR14420-1GEA 0 EUR
APOSOR14420-1G
4-(1,3-Dithiolan-2-yl)-2-methoxy-6-nitrophenol
4-(1,3-Dithiolan-2-yl)-2-methoxy-6-nitrophenol
Kaava: C₁₀H₁₁NO₄S₂
MW: 273,329 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00169565
CAS-numero: 331460-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR