Ethyl 4-(5-methyl-1,2,4-oxadiazol-3-yl)benzoate

Toimittaja: Apollo Scientific

APOSOR1449-1GEA 103 EUR
APOSOR1449-1G
Ethyl 4-(5-methyl-1,2,4-oxadiazol-3-yl)benzoate
Ethyl 4-(5-methyl-1,2,4-oxadiazol-3-yl)benzoate
Kaava: C₁₂H₁₂N₂O₃
MW: 232,235 g/mol
Sulamispiste: 103,2…103.5 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD07368532
CAS-numero: 850375-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR