5-(Bromoacetyl)-1-methyl-1H-benzimidazole hydrobromide 97%

Varoitus

APOSOR15382-1GEA 0 EUR
APOSOR15382-1G APOSOR15382-250MG
5-(Bromoacetyl)-1-methyl-1H-benzimidazole hydrobromide 97%
5-(Bromoacetyl)-1-methyl-1H-benzimidazole hydrobromide
Kaava: C₁₀H₉BrN₂O·BrH
MW: 334,007 g/mol
Sulamispiste: 196…197 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09817461
CAS-numero: 944450-78-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR