5-Bromo-4-methyl-1H-benzimidazole 97%

APOSOR19568-100MGEA 0 EUR
APOSOR19568-100MG
5-Bromo-4-methyl-1H-benzimidazole 97%
5-Bromo-4-methyl-1H-benzimidazole
Kaava: C₈H₇BrN₂
MW: 211,059 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD18642426
CAS-numero: 952511-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR