2-(1,3-Benzodioxol-5-yl)indolizine-3-carboxaldehyde

APOSOR1972-1GEA 0 EUR
APOSOR1972-1G
2-(1,3-Benzodioxol-5-yl)indolizine-3-carboxaldehyde
2-(1,3-Benzodioxol-5-yl)indolizine-3-carboxaldehyde
Kaava: C₁₆H₁₁NO₃
MW: 265,263 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD08056205
CAS-numero: 893612-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR