2-(4-Chlorophenyl)indolizine-3-carboxaldehyde

APOSOR1976-1GEA 0 EUR
APOSOR1976-1G APOSOR1976-500MG
2-(4-Chlorophenyl)indolizine-3-carboxaldehyde
2-(4-Chlorophenyl)indolizine-3-carboxaldehyde
Kaava: C₁₅H₁₀ClNO
MW: 255,699 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD05178248
CAS-numero: 558424-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR