3-Amino-2-(biphenyl-2-ylamino)benzoic acid

Varoitus

APOSOR1983-1GEA 0 EUR
APOSOR1983-1G APOSOR1983-500MG
3-Amino-2-(biphenyl-2-ylamino)benzoic acid
3-Amino-2-(biphenyl-2-ylamino)benzoic acid
Kaava: C₁₉H₁₆N₂O₂
MW: 304,343 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD08056213
CAS-numero: 893613-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR