2-(3-Chlorophenyl)-4-[(methylamino)methyl]-1,3-thiazole 90%

Vaara

APOSOR2067-1GEA 0 EUR
APOSOR2067-1G
2-(3-Chlorophenyl)-4-[(methylamino)methyl]-1,3-thiazole 90%
2-(3-Chlorophenyl)-4-[(methylamino)methyl]-1,3-thiazole
Kaava: C₁₁H₁₁ClN₂S
MW: 238,736 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD08690268
CAS-numero: 864068-99-5
UN: 2922
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR