2-(4-Chlorophenoxy)pyridine-3-carbonyl chloride

APOSOR21706-1GEA 0 EUR
APOSOR21706-1G
2-(4-Chlorophenoxy)pyridine-3-carbonyl chloride
2-(4-Chlorophenoxy)pyridine-3-carbonyl chloride
Kaava: C₁₂H₇Cl₂NO₂
MW: 268,095 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00067837
CAS-numero: 51362-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR