3-Amino-2-benzylpropanoic acid, N-FMOC protected 99%

Toimittaja: Apollo Scientific

APOSOR2226-250MGEA 324 EUR
APOSOR2226-250MG
3-Amino-2-benzylpropanoic acid, N-FMOC protected 99%
3-Amino-2-benzylpropanoic acid, N-FMOC protected
Kaava: C₂₅H₂₃NO₄
MW: 401,454 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02258442
CAS-numero: 683217-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR