1-Aminocycloheptanecarboxylic acid, N-BOC protected 97%

Varoitus

APOSOR2230-250MGEA 0 EUR
APOSOR2230-250MG APOSOR2230-1G
1-Aminocycloheptanecarboxylic acid, N-BOC protected 97%
1-Aminocycloheptanecarboxylic acid, N-BOC protected
Kaava: C₁₃H₂₃NO₄
MW: 257,326 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03426130
CAS-numero: 199330-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR