2,6-Dimethylundecane

Varoitus

APOSOR2377-5MLEA 0 EUR
APOSOR2377-5ML APOSOR2377-1ML
2,6-Dimethylundecane
2,6-Dimethylundecane
Kaava: C₁₃H₂₈
MW: 184,361 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00048876
CAS-numero: 17301-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR