Bicyclo[2.2.1]hept-5-en-2-yl isothiocyanate

Varoitus

APOSOR24481-250MGEA 0 EUR
APOSOR24481-250MG APOSOR24481-1G
Bicyclo[2.2.1]hept-5-en-2-yl isothiocyanate
Bicyclo[2.2.1]hept-5-en-2-yl isothiocyanate
Kaava: C₈H₉NS
MW: 151,229 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00122413
CAS-numero: 92819-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR