6-Hydrazino-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine 97%

Vaara

APOSOR25210-5GEA 0 EUR
APOSOR25210-5G APOSOR25210-1G
6-Hydrazino-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine 97%
6-Hydrazino-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine
Kaava: C₉H₁₃N₅
MW: 191,233 g/mol
Sulamispiste: 199…205 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00067969
CAS-numero: 175202-00-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR