(4-Aminopiperidin-4-yl)acetic acid, N1-BOC 4-FMOC protected 98%

Toimittaja: Apollo Scientific

APOSOR2629-250MGEA 462 EUR
APOSOR2629-250MG
(4-Aminopiperidin-4-yl)acetic acid, N1-BOC 4-FMOC protected 98%
(4-Aminopiperidin-4-yl)acetic acid, N1-BOC 4-FMOC protected
Kaava: C₂₇H₃₂N₂O₆
MW: 480,553 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD08457839
CAS-numero: 946682-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR