Ethyl 2,5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate

Varoitus

APOSOR29457-5GEA 0 EUR
APOSOR29457-5G
Ethyl 2,5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate
Ethyl 2,5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate
Kaava: C₁₅H₁₇NO₂
MW: 243,301 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00204227
CAS-numero: 76546-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR