4-(Dichloromethylene)morpholin-4-ium

APOSOR300166-1GEA 0 EUR
APOSOR300166-1G
4-(Dichloromethylene)morpholin-4-ium
4-(Dichloromethylene)morpholin-4-ium
Storage Temperature: Huoneenlämpö
MDL numero: MFCD19439686
CAS-numero: 790600-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR