Ethyl 3-amino-3-phenylpropionate 95+%

Toimittaja: Apollo Scientific

APOSOR300757-250MGEA 95.1 EUR
APOSOR300757-250MG APOSOR300757-1G
Ethyl 3-amino-3-phenylpropionate 95+%
Ethyl 3-amino-3-phenylpropionate
Kaava: C₁₁H₁₅NO₂
MW: 193,242 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00749016
CAS-numero: 6335-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR