4-Oxopiperidine-3-carboxamide

APOSOR302005-500MGEA 0 EUR
APOSOR302005-500MG
4-Oxopiperidine-3-carboxamide
4-Oxopiperidine-3-carboxamide
Kaava: C₆H₁₀N₂O₂
MW: 142,156 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD22690731
CAS-numero: 23608-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR