5,6-Dihydro-2-iodoimidazo[1,2-d][1,4]benzoxazepine-10-carboxylic acid, methyl ester

APOSOR302110-1GEA 0 EUR
APOSOR302110-1G
5,6-Dihydro-2-iodoimidazo[1,2-d][1,4]benzoxazepine-10-carboxylic acid, methyl ester
5,6-Dihydro-2-iodoimidazo[1,2-d][1,4]benzoxazepine-10-carboxylic acid, methyl ester
Kaava: C₁₃H₁₁IN₂O₃
MW: 370,143 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD22581525
CAS-numero: 1282516-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR