Methyl 3-(dimethoxymethyl)picolinate

APOSOR302201-1GEA 0 EUR
APOSOR302201-1G
Methyl 3-(dimethoxymethyl)picolinate
Methyl 3-(dimethoxymethyl)picolinate
Kaava: C₁₀H₁₃NO₄
MW: 211,215 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD22041932
CAS-numero: 133155-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR