3-Cyclopentyl-4-oxobutanenitrile

APOSOR302340-1GEA 0 EUR
APOSOR302340-1G
3-Cyclopentyl-4-oxobutanenitrile
3-Cyclopentyl-4-oxobutanenitrile
Kaava: C₉H₁₃NO
MW: 151,206 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD22690781
CAS-numero: 1439823-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR