tert-Butyl (2-((2,4-dichlorobenzyl)amino)ethyl)carbamate

APOSOR302348-1GEA 681 EUR
APOSOR302348-1G
tert-Butyl (2-((2,4-dichlorobenzyl)amino)ethyl)carbamate
tert-Butyl (2-((2,4-dichlorobenzyl)amino)ethyl)carbamate
Kaava: C₁₄H₂₀Cl₂N₂O₂
MW: 319,227 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD22690787
CAS-numero: 1439823-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR