Methyl 2-chloro-4-morpholinopyrido[2,3-d]pyrimidine-7-carboxylate

Toimittaja: Apollo Scientific

APOSOR302469-1GEA 729 EUR
APOSOR302469-1G
Methyl 2-chloro-4-morpholinopyrido[2,3-d]pyrimidine-7-carboxylate
Methyl 2-chloro-4-morpholinopyrido[2,3-d]pyrimidine-7-carboxylate
Kaava: C₁₃H₁₃ClN₄O₃
MW: 308,72 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD21648263
CAS-numero: 1227958-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR