5-(Morpholinosulphonyl)pyridin-2-amine

APOSOR302544-1GEA 0 EUR
APOSOR302544-1G
5-(Morpholinosulphonyl)pyridin-2-amine
5-(Morpholinosulphonyl)pyridin-2-amine
Kaava: C₉H₁₃N₃O₃S
MW: 243,283 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD06307828
CAS-numero: 627840-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR