2-Methyl-6-morpholinopyrimidin-4-amine

APOSOR302561-1GEA 0 EUR
APOSOR302561-1G
2-Methyl-6-morpholinopyrimidin-4-amine
2-Methyl-6-morpholinopyrimidin-4-amine
Kaava: C₉H₁₄N₄O
MW: 194,234 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09880706
CAS-numero: 28732-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR