(2-Oxo-1,2-dihydropyridin-3-yl)boronic acid

Varoitus

APOSOR302583-250MGEA 0 EUR
APOSOR302583-250MG APOSOR302583-100MG
(2-Oxo-1,2-dihydropyridin-3-yl)boronic acid
(2-Oxo-1,2-dihydropyridin-3-yl)boronic acid
Kaava: C₅H₆BNO₃
MW: 138,917 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD12964572
CAS-numero: 951655-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR