Ethyl 5-tert-butyl-1,3-oxazole-4-carboxylate

Toimittaja: Apollo Scientific

APOSOR303146-1GEA 184 EUR
APOSOR303146-1G APOSOR303146-500MG
Ethyl 5-tert-butyl-1,3-oxazole-4-carboxylate
Ethyl 5-tert-butyl-1,3-oxazole-4-carboxylate
Kaava: C₁₀H₁₅NO₃
MW: 197,231 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00105461
CAS-numero: 714273-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR