1-[5-(Benzyloxy)-2-hydroxy-3-nitrophenyl]ethanone

APOSOR305016-250MGEA 0 EUR
APOSOR305016-250MG APOSOR305016-1G
1-[5-(Benzyloxy)-2-hydroxy-3-nitrophenyl]ethanone
1-[5-(Benzyloxy)-2-hydroxy-3-nitrophenyl]ethanone
Kaava: C₁₅H₁₃NO₅
MW: 287,267 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD17169905
CAS-numero: 861841-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR