tert-Butyl 3-iodo-5-nitro-1H-indazole-1-carboxylate

APOSOR305066-250MGEA 0 EUR
APOSOR305066-250MG
tert-Butyl 3-iodo-5-nitro-1H-indazole-1-carboxylate
tert-Butyl 3-iodo-5-nitro-1H-indazole-1-carboxylate
Kaava: C₁₂H₁₂IN₃O₄
MW: 389,146 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD11110274
CAS-numero: 459133-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR