3-Amino-1H-indazol-4-ol

APOSOR305096-250MGEA 0 EUR
APOSOR305096-250MG
3-Amino-1H-indazol-4-ol
3-Amino-1H-indazol-4-ol
Kaava: C₇H₇N₃O
MW: 149,15 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD13176941
CAS-numero: 88805-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR