Methyl 5-bromo-1H-indazole-4-carboxylate

Toimittaja: Apollo Scientific

APOSOR305114-250MGEA 373 EUR
APOSOR305114-250MG
Methyl 5-bromo-1H-indazole-4-carboxylate
Methyl 5-bromo-1H-indazole-4-carboxylate
Kaava: C₉H₇BrN₂O₂
MW: 255,068 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD13177017
CAS-numero: 1037840-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR