6-Bromo-1H-indazol-4-ol

APOSOR305140-250MGEA 0 EUR
APOSOR305140-250MG
6-Bromo-1H-indazol-4-ol
6-Bromo-1H-indazol-4-ol
Kaava: C₇H₅BrN₂O
MW: 213,031 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD07781339
CAS-numero: 885518-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR