5-Ethoxyimidazo[1,2-a]pyridine

APOSOR305162-250MGEA 0 EUR
APOSOR305162-250MG
5-Ethoxyimidazo[1,2-a]pyridine
5-Ethoxyimidazo[1,2-a]pyridine
Kaava: C₉H₁₀N₂O
MW: 162,189 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD13177215
CAS-numero: 3323-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR