Methyl (2E)-3-(1H-indol-7-yl)prop-2-enoate

Toimittaja: Apollo Scientific

APOSOR305243-250MGEA 359 EUR
APOSOR305243-250MG
Methyl (2E)-3-(1H-indol-7-yl)prop-2-enoate
Methyl (2E)-3-(1H-indol-7-yl)prop-2-enoate
Kaava: C₁₂H₁₁NO₂
MW: 201,221 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD13177665
CAS-numero: 104682-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR