Ethyl (2,3-dihydro-1H-indol-4-yloxy)acetate

Toimittaja: Apollo Scientific

APOSOR305257-1GEA 534 EUR
APOSOR305257-1G APOSOR305257-250MG
Ethyl (2,3-dihydro-1H-indol-4-yloxy)acetate
Ethyl (2,3-dihydro-1H-indol-4-yloxy)acetate
Kaava: C₁₂H₁₅NO₃
MW: 221,252 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD11869058
CAS-numero: 947382-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR