6-Bromonaphthalene-1-carboxylic acid

APOSOR305312-250MGEA 0 EUR
APOSOR305312-250MG
6-Bromonaphthalene-1-carboxylic acid
6-Bromonaphthalene-1-carboxylic acid
Kaava: C₁₁H₇BrO₂
MW: 251,076 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09842464
CAS-numero: 51934-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR