2,3-Dihydro-1H-indol-5-ylboronic acid

APOSOR305603-250MGEA 0 EUR
APOSOR305603-250MG
2,3-Dihydro-1H-indol-5-ylboronic acid
2,3-Dihydro-1H-indol-5-ylboronic acid
Kaava: C₈H₁₀BNO₂
MW: 162,982 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD26743830
CAS-numero: 935853-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR