4-Cyclopropyl-1H-pyrazol-3-amine

Varoitus

APOSOR306666-1GEA 0 EUR
APOSOR306666-1G
4-Cyclopropyl-1H-pyrazol-3-amine
4-Cyclopropyl-1H-pyrazol-3-amine
Kaava: C₆H₉N₃
MW: 123,156 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD08752485
CAS-numero: 673475-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR