2-(3-Ethoxy-benzyl)-piperidine hydrochloride

Toimittaja: Apollo Scientific

APOSOR307739-1GEA 187 EUR
APOSOR307739-1G
2-(3-Ethoxy-benzyl)-piperidine hydrochloride
2-(3-Ethoxy-benzyl)-piperidine hydrochloride
Kaava: C₁₄H₂₁NO·ClH
MW: 255,784 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD08669727
CAS-numero: 1170163-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR