2-(4-Ethoxy-benzyl)-piperidine hydrochloride

Toimittaja: Apollo Scientific

APOSOR307744-1GEA 437 EUR
APOSOR307744-1G
2-(4-Ethoxy-benzyl)-piperidine hydrochloride
2-(4-Ethoxy-benzyl)-piperidine hydrochloride
Kaava: C₁₄H₂₁NO·ClH
MW: 255,784 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD08669738
CAS-numero: 1172745-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR