3,4-Dihydro-4-methyl-2H-1,4-benzoxazine-2-carboxylic acid 97%

Varoitus

APOSOR30809-1GEA 0 EUR
APOSOR30809-1G APOSOR30809-250MG
3,4-Dihydro-4-methyl-2H-1,4-benzoxazine-2-carboxylic acid 97%
3,4-Dihydro-4-methyl-2H-1,4-benzoxazine-2-carboxylic acid
Kaava: C₁₀H₁₁NO₃
MW: 193,199 g/mol
Sulamispiste: 143…153 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD11506351
CAS-numero: 212578-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR