5-(Bromomethyl)-1H-indazole hydrobromide

Varoitus

APOSOR30832-1GEA 0 EUR
APOSOR30832-1G
5-(Bromomethyl)-1H-indazole hydrobromide
5-(Bromomethyl)-1H-indazole hydrobromide
Kaava: C₈H₇BrN₂·BrH
MW: 291,971 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD11111372
CAS-numero: 192369-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR