2-Amino-3-methylquinoline hydrochloride

APOSOR309451-1GEA 0 EUR
APOSOR309451-1G
2-Amino-3-methylquinoline hydrochloride
2-Amino-3-methylquinoline hydrochloride
Kaava: C₁₀H₁₀N₂·ClH
MW: 194,661 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD07644600
CAS-numero: 1172338-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR