tert-Butyl(2-oxo-3-phenoxypropyl)amine hydrochloride

APOSOR310339-1GEA 0 EUR
APOSOR310339-1G APOSOR310339-5G
tert-Butyl(2-oxo-3-phenoxypropyl)amine hydrochloride
tert-Butyl(2-oxo-3-phenoxypropyl)amine hydrochloride
Kaava: C₁₃H₁₉NO₂·ClH
MW: 257,756 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD26523313
CAS-numero: 1565845-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR