2-(1,6-Naphthyridin-2-ylformamido)acetamide

APOSOR310439-1GEA 0 EUR
APOSOR310439-1G
2-(1,6-Naphthyridin-2-ylformamido)acetamide
2-(1,6-Naphthyridin-2-ylformamido)acetamide
Kaava: C₁₁H₁₀N₄O₂
MW: 230,223 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD27957300
CAS-numero: 1565845-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR