9H-Fluoren-9-ylmethyl N-[(2S)-4-(methylsulphanyl)-1-oxobutan-2-yl]carbamate

APOSOR310441-1GEA 0 EUR
APOSOR310441-1G
9H-Fluoren-9-ylmethyl N-[(2S)-4-(methylsulphanyl)-1-oxobutan-2-yl]carbamate
9H-Fluoren-9-ylmethyl N-[(2S)-4-(methylsulphanyl)-1-oxobutan-2-yl]carbamate
Kaava: C₂₀H₂₁NO₃S
MW: 355,451 g/mol
Sulamispiste: 115 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD27957303
CAS-numero: 211929-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR