Methyl 2-methyl-3-(pyrazin-2-yl)-3-oxopropionate

APOSOR310604-1GEA 0 EUR
APOSOR310604-1G
Methyl 2-methyl-3-(pyrazin-2-yl)-3-oxopropionate
Methyl 2-methyl-3-(pyrazin-2-yl)-3-oxopropionate
Kaava: C₉H₁₀N₂O₃
MW: 194,187 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD06245585
CAS-numero: 324737-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR